missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentylacetic acid, 97+%

Manufacturer:  Alfa Aesar™ A1569622

 View more versions of this product

Catalog No. AAA1569622



Specifications

Specifications

Cyclopentylacetic acid
Rancid
12°C to 14°C
1123-00-8
MFCD00001387
cyclopentylacetic acid, cyclopentaneacetic acid, cycylopentylacetic acid, carboxymethyl cyclopentane, cyclopentyl acetic acid, cyclopentyacetic acid, cyclopentanacetic acid, cyclopentyl-acetic acid, 2-cyclopentylaceticacid, cyclopentane acetic acid
C1CCC(C1)CC(=O)O
128.171
128.17
114°C (237°F)
231°C
100g
C7H12O2
2040821
YVHAIVPPUIZFBA-UHFFFAOYSA-N
2-cyclopentylacetic acid
71606
≥97%

Chemical Identifiers

1123-00-8
128.171
YVHAIVPPUIZFBA-UHFFFAOYSA-N
71606
C1CCC(C1)CC(=O)O
C7H12O2
MFCD00001387
cyclopentylacetic acid, cyclopentaneacetic acid, cycylopentylacetic acid, carboxymethyl cyclopentane, cyclopentyl acetic acid, cyclopentyacetic acid, cyclopentanacetic acid, cyclopentyl-acetic acid, 2-cyclopentylaceticacid, cyclopentane acetic acid
2-cyclopentylacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents