Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentylacetic acid, 97+%

$82.04 - $526.43

Chemical Identifiers

Chemical Name or Material Cyclopentylacetic acid
CAS 1123-00-8
Molecular Formula C7H12O2
Formula Weight 128.17
MDL Number MFCD00001387
Beilstein 2040821
Synonym Cyclopentaneacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1569606

 
alfa aesar™
A1569606
5g Each for $82.04

AAA1569614

 
alfa aesar™
A1569614
25g Each for $167.78

AAA1569622

 
alfa aesar™
A1569622
100g Each for $526.43
Description & Specifications

Specifications

Formula Weight 128.17
Assay ≥97%
Boiling Point 231°C
Chemical Name or Material Cyclopentylacetic acid
Molecular Formula C7H12O2
MDL Number MFCD00001387
CAS 1123-00-8
Beilstein 2040821
Synonym Cyclopentaneacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 114°C (237°F)
Odor Rancid
Melting Point 12° to 14°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.