Please login to your online account to display your discounted pricing

Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC397251000

 View more versions of this product

Catalog No. AC397251000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.4%
Physical Form Liquid
Formula Weight 100.16
Quantity 100mL
Color Clear
Merck Index 15, 2742
Packaging AcroSeal™ Glass Bottle
Purity Grade Extra Dry
Density 0.8600g/mL
UN Number 1993
Chemical Name or Material Cyclopentyl methyl ether
Residue after Evaporation 0.005% max.
Molecular Formula C6H12O
Linear Formula C5H9OCH3
MDL Number MFCD08276401
CAS 5614-37-9
Name Note Stabilized, Extra Dry
Assay Percent Range 99.5%
Stabilizer 50ppm (BHT) approximately
Water 0.005% max.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclopentyl methyl ether
Name Note Stabilized, Extra Dry
CAS 5614-37-9
Purity Grade Extra Dry
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C6H12O
Linear Formula C5H9OCH3
Formula Weight 100.16
MDL Number MFCD08276401
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents