missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ A1174503

 View more versions of this product

Catalog No. AAA1174503



Description

Description

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Soluble in ethanol, acetone and hydrolyzes with water.

Notes
Moisture Sensitive, store away form water/moisture. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store under dry inert gas.
Specifications

Specifications

Cyclopentyl isothiocyanate
1g
C6H9NS
UN2922
cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
C1CCC(C1)N=C=S
127.205
127.21
75°C to 76°C (10mmHg)
33522-03-1
MFCD00040877
2715701
PJOODZCPFADLCI-UHFFFAOYSA-N
isothiocyanatocyclopentane
141792
98%

Chemical Identifiers

33522-03-1
127.205
PJOODZCPFADLCI-UHFFFAOYSA-N
141792
C1CCC(C1)N=C=S
C6H9NS
MFCD00040877
cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
isothiocyanatocyclopentane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

SDS Documents