Learn More
Cyclopentanone, 99+%, pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 111530100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopentanone | |
0.9500g/mL | |
26°C | |
99% min. (GC) | |
C5H8O | |
C5H8(=O) | |
10L | |
0.95 | |
adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy | |
BGTOWKSIORTVQH-UHFFFAOYSA-N | |
cyclopentanone | |
8452 | |
CHEBI:16486 | |
99+% |
120-92-3 | |
130.0°C to 131.0°C | |
Authentic | |
Plastic drum | |
1.4360 to 1.4380 | |
MFCD00001409 | |
07,5 | |
15,2738 | |
Solubility in water: 301g/L (20°C). Other solubilities: miscible with alcohol,acetone and ether | |
O=C1CCCC1 | |
84.12 | |
0.8 mPa.s (20°C) | |
84.12 | |
Pure |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-435-9
RUO â Research Use Only