Learn More
Cyclopentanethiol, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC435690250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclopentanethiol | |
Colorless to Yellow | |
25°C | |
Authentic | |
MFCD00001369 | |
25g | |
SC1CCCC1 | |
102.20 | |
102.2 | |
98% |
1679-07-8 | |
129°C to 131°C | |
97.5% min. (GC) | |
C5H10S | |
acmc-209dxb, cyclopen-tanethiol, cyclopentane thiol, cyclopentanethiol, cyclopentyl mercaptan, cyclopentyl thiol, cyclopentylmercaptan, cyclopentylthiol, fema no. 3262, mercaptocyclopentane | |
WVDYBOADDMMFIY-UHFFFAOYSA-N | |
cyclopentanethiol | |
15510 | |
Liquid |
Chemical Identifiers
1679-07-8 | |
102.20 | |
WVDYBOADDMMFIY-UHFFFAOYSA-N | |
15510 | |
SC1CCCC1 |
C5H10S | |
MFCD00001369 | |
acmc-209dxb, cyclopen-tanethiol, cyclopentane thiol, cyclopentanethiol, cyclopentyl mercaptan, cyclopentyl thiol, cyclopentylmercaptan, cyclopentylthiol, fema no. 3262, mercaptocyclopentane | |
cyclopentanethiol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
EINECSNumber : 216-841-3
RUO – Research Use Only