Learn More
Cyclopentane, HPLC Grade, Thermo Scientific Chemicals
For HPLC and Spectrophotometry
Supplier: Thermo Scientific Chemicals 044814AK
Description
Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclopentane | |
287-92-3 | |
Colorless | |
47°C to 49°C | |
380°C (716°F) | |
C5H10 | |
MFCD00001356 | |
1900195 | |
Miscible with ethanol,ether and acetone. Slightly miscible with water. | |
C1CCCC1 | |
70.135 | |
CHEBI:23492 | |
398mm Hg at 20°C | |
Liquid |
Contains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons . | |
°C to 95°C | |
0.748 g/cm3 at 20°C | |
-37°C (-34°F) | |
Petrol-like | |
1.406 | |
250 mL | |
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
cyclopentane | |
9253 | |
70.14 | |
HPLC |
Chemical Identifiers
287-92-3 | |
70.135 | |
RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
9253 | |
cyclopentane |
C5H10 | |
MFCD00001356 | |
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
CHEBI:23492 | |
C1CCCC1 |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
H225-H304-H335-H336
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 206-016-6
RTECSNumber : GY2390000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only