missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentane, HPLC Grade, Alfa Aesar™

For HPLC and Spectrophotometry

Manufacturer:  Alfa Aesar 44814K2

 View more versions of this product

Catalog No. AA44814K2



Specifications

Specifications

Cyclopentane
Petrol-like
Colorless
Contains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons .
C5H10
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
C1CCCC1
70.135
CHEBI:23492
Liquid
380°C (716°F)
398mm Hg at 20°C
1L
287-92-3
MFCD00001356
RGSFGYAAUTVSQA-UHFFFAOYSA-N
cyclopentane
9253
70.14
HPLC

Chemical Identifiers

287-92-3
70.135
RGSFGYAAUTVSQA-UHFFFAOYSA-N
9253
cyclopentane
C5H10
MFCD00001356
pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
CHEBI:23492
C1CCCC1
Safety and Handling

Safety and Handling

H225-H412: Highly flammable liquid and vapour. Harmful to aquatic life with long lasting effects.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 206-016-6

SDS Documents