missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 1270-98-0 |
---|---|
Molecular Formula | C5H5Cl3Ti |
Molecular Weight (g/mol) | 219.36 |
MDL Number | MFCD00003724 |
InChI Key | QOXHZZQZTIGPEV-UHFFFAOYSA-K |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC215770025
|
Thermo Scientific Chemicals
215770025 |
2.5 g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC215770100
|
Thermo Scientific Chemicals
215770100 |
10 g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1270-98-0 | |
219.36 | |
QOXHZZQZTIGPEV-UHFFFAOYSA-K |
C5H5Cl3Ti | |
MFCD00003724 |
Specifications
1270-98-0 | |
99% | |
MFCD00003724 | |
QOXHZZQZTIGPEV-UHFFFAOYSA-K | |
219.36 | |
Authentic | |
Cyclopentadienyltitanium trichloride, 99% |
2.5 g | |
C5H5Cl3Ti | |
Solubility in water: reacts | |
219.36 | |
99% | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RTECSNumber : XR1927300
RUO â Research Use Only