missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentadienyltitanium trichloride

Manufacturer:  Alfa Aesar™ 7713406

 View more versions of this product

Catalog No. AA7713406



Specifications

Specifications

Cyclopentadienyltitanium trichloride
C5H5Cl3Ti
UN3261
QOXHZZQZTIGPEV-UHFFFAOYSA-K
219.35
Moisture Sensitive
210°C (decomposition)
1270-98-0
MFCD00003724
Trichlorocyclopentadienyltitanium
219.31
Crystalline
Yellow to Orange
5g

Chemical Identifiers

1270-98-0
219.31
QOXHZZQZTIGPEV-UHFFFAOYSA-K
C5H5Cl3Ti
MFCD00003724
Trichlorocyclopentadienyltitanium
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : XR1927300

SDS Documents