Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentadienylrhenium tricarbonyl, 99%

Chemical Identifiers

Chemical Name or Material Cyclopentadienylrhenium tricarbonyl
CAS 12079-73-1
Formula Weight 335.33
MDL Number MFCD00058885
UN Number UN3466
Synonym Tricarbonylcyclopentadienylrhenium
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3365502

  SDS
alfa aesar™
3365502
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3365504

  SDS
alfa aesar™
3365504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 335.33
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Color White
Chemical Name or Material Cyclopentadienylrhenium tricarbonyl
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
CAS 12079-73-1
Health Hazard 1 Danger
Quantity 0.5g
Physical Form Powder
UN Number UN3466
Synonym Tricarbonylcyclopentadienylrhenium
MDL Number MFCD00058885
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.