Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentadienylmanganese tricarbonyl

Chemical Identifiers

Chemical Name or Material Cyclopentadienylmanganese tricarbonyl
CAS 12079-65-1
Formula Weight 204.06
MDL Number MFCD00001440
UN Number UN3466
Synonym Tricarbonylcyclopentadienylmanganese
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3860603

  SDS
alfa aesar™
3860603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3860606

  SDS
alfa aesar™
3860606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3860614

  SDS
alfa aesar™
3860614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.06
Chemical Name or Material Cyclopentadienylmanganese tricarbonyl
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00001440
CAS 12079-65-1
UN Number UN3466
Synonym Tricarbonylcyclopentadienylmanganese
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Sensitivity Air Sensitive
Melting Point 77°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.