missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclopentadienyliron dicarbonyl dimer, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC209301000

 View more versions of this product

Catalog No. AC209301000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclopentadienyliron dicarbonyl dimer
Authentic
Brown to Purple-Red
100g
99%
MFCD00001436
Crystalline Powder
98.5% min.
Glass bottle
194.0°C
12154-95-9
C14H10Fe2O4
353.92
99%

Chemical Identifiers

12154-95-9
C14H10Fe2O4
MFCD00001436
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents