Please login to your online account to display your discounted pricing

Cyclopentadienyliron dicarbonyl dimer, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyclopentadienyliron dicarbonyl dimer
Assay Percent Range 99%
Molecular Formula C14H10Fe2O4
Formula Weight 353.92
MDL Number MFCD00001436
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC209301000

  SDS
thermo fisher scientific
AC209301000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 353.92
Quantity 100g
Color Red-purple to very dark brown
Chemical Name or Material Cyclopentadienyliron dicarbonyl dimer
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C14H10Fe2O4
MDL Number MFCD00001436
CAS 12154-95-9
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.