Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentadecanone, 99%

$19.73 - $83.77

Chemical Identifiers

Chemical Name or Material Cyclopentadecanone
CAS 502-72-7
Molecular Formula C15H28O
Formula Weight 224.39
MDL Number MFCD00001266
Beilstein 1618444
Synonym Exaltone; Normuscone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1765206

  SDS
alfa aesar™
A1765206
5g Each for $83.77

AAA1765203

  SDS
alfa aesar™
A1765203
1g Each for $19.73
Description & Specifications

Specifications

Formula Weight 224.39
Chemical Name or Material Cyclopentadecanone
TSCA TSCA
Assay 99%
Molecular Formula C15H28O
MDL Number MFCD00001266
CAS 502-72-7
Beilstein 1618444
Synonym Exaltone; Normuscone
RTECS Number GY0900000
Boiling Point 120°C (0.3mmHg)
Melting Point 62° to 66°C
Quantity 5g