Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclopentadecanol, 98%

$59.38 - $232.04

Chemical Identifiers

Chemical Name or Material Cyclopentadecanol
CAS 4727-17-7
Molecular Formula C15H30O
Formula Weight 226.4
MDL Number MFCD00039425
Synonym Hydroxycyclopentadecane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1888803

  SDS
alfa aesar™
A1888803
1g Each for $59.38

AAA1888806

  SDS
alfa aesar™
A1888806
5g Each for $232.04
Description & Specifications

Specifications

Formula Weight 226.4
Chemical Name or Material Cyclopentadecanol
TSCA TSCA
Assay 98%
Molecular Formula C15H30O
MDL Number MFCD00039425
CAS 4727-17-7
Synonym Hydroxycyclopentadecane
Quantity 1g