Learn More
Cyclooctylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 111460250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclooctylamine | |
-48°C | |
0.9200g/mL | |
62°C | |
96% min. (GC) | |
C8H17N | |
C8H15NH2 | |
25 g | |
0.92 | |
Solubility in water: 10.2g/L (20°C) | |
C1CCCC(CCC1)N | |
127.23 | |
5 mPa.s (20°C) | |
97% |
5452-37-9 | |
Colorless to Yellow | |
190°C | |
Authentic | |
Glass bottle | |
1.4800 to 1.482 | |
MFCD00001748 | |
12, IV, 128 | |
cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc | |
HSOHBWMXECKEKV-UHFFFAOYSA-N | |
cyclooctanamine | |
2903 | |
127.23 | |
Liquid |
Chemical Identifiers
5452-37-9 | |
127.23 | |
HSOHBWMXECKEKV-UHFFFAOYSA-N | |
2903 | |
C1CCCC(CCC1)N |
C8H17N | |
MFCD00001748 | |
cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc | |
cyclooctanamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 226-694-7
RUO – Research Use Only