Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclooctylamine, 97+%

Chemical Identifiers

Chemical Name or Material Cyclooctylamine
CAS 5452-37-9
Molecular Formula C8H17N
Formula Weight 127.23
MDL Number MFCD00001748
UN Number UN2735
Beilstein 2689120
Synonym Aminocyclooctane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0098106

  SDS
alfa aesar™
L0098106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0098114

  SDS
alfa aesar™
L0098114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.23
Chemical Name or Material Cyclooctylamine
Molecular Formula C8H17N
TSCA TSCA
Assay ≥97%
MDL Number MFCD00001748
CAS 5452-37-9
UN Number UN2735
Beilstein 2689120
Synonym Aminocyclooctane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 62°C (143°F)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Odor Amine-like
Boiling Point 189° to 190°C
Melting Point −48°C
Quantity 5g

Applications
Cyclooctylamine is used as pharmaceutical intermediates.

Solubility
Soluble in water. 10.2 g/L at 20°C.

Notes
Protect from moisture, heat. Avoid excessive and light exposure. Air sensitive. Store at +15°C to +25°C.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.