Please login to your online account to display your discounted pricing

Alfa Aesar™ (Cyclooctatetraene)iron tricarbonyl, 98%

Chemical Identifiers

Chemical Name or Material (Cyclooctatetraene)iron tricarbonyl
CAS 12093-05-9
Formula Weight 244.03
MDL Number MFCD00001755
UN Number UN3466
Synonym Tricarbonylcyclooctatetraene iron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3940202

  SDS
alfa aesar™
3940202
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3940204

  SDS
alfa aesar™
3940204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.03
Health Hazard 1 Warning
Assay 98%
Chemical Name or Material (Cyclooctatetraene)iron tricarbonyl
UN Number UN3466
Physical Form Crystalline
Synonym Tricarbonylcyclooctatetraene iron
Quantity 0.5g
Sensitivity Air Sensitive
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Melting Point 93° to 95°C
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00001755
Color Red-brown
CAS 12093-05-9
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.