Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyltrichlorosilane, 97+%

Chemical Identifiers

Chemical Name or Material Cyclohexyltrichlorosilane
CAS 98-12-4
Molecular Formula C6H11Cl3Si
Formula Weight 217.60g/mol
MDL Number MFCD00021280
UN Number UN1763
Beilstein 2935257
Synonym Trichlorocyclohexylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2116909

 
alfa aesar™
B21169-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Acrid
Boiling Point 90° to 91°C (10mmHg)
Quantity 10g
Flash Point 79°C (174°F)
Chemical Name or Material Cyclohexyltrichlorosilane
CAS 98-12-4
Assay ≥97%
Molecular Formula C6H11Cl3Si
Formula Weight 217.60g/mol
MDL Number MFCD00021280
UN Number UN1763
Beilstein 2935257
Synonym Trichlorocyclohexylsilane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CYCLOHEXYLTRICHLOROSILANE
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.