Learn More
Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H51165AE
Description
Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
Specifications
Cyclohexylmagnesium chloride | |
C6H11ClMg | |
100 mL | |
Air and moisture sensitive | |
It reacts violently with water. | |
Cl[Mg]C1CCCCC1 | |
142.91 | |
142.91 |
931-51-1 | |
MFCD00003816 | |
UN2924 | |
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride | |
WMJMABVHDMRMJA-UHFFFAOYSA-M | |
magnesium;cyclohexane;chloride | |
13605 | |
1M in MeTHF |
Chemical Identifiers
931-51-1 | |
142.91 | |
WMJMABVHDMRMJA-UHFFFAOYSA-M | |
13605 | |
Cl[Mg]C1CCCCC1 |
C6H11ClMg | |
MFCD00003816 | |
cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride | |
magnesium;cyclohexane;chloride |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only