missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexylidenecyanoacetic acid, 97%

Chemical Identifiers

CAS 37107-50-9
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00013799
InChI Key KMAMEGWTNNOKOE-UHFFFAOYSA-N
Synonym cyclohexylidenecyanoacetic acid, cyanocyclohexylideneacetic acid, cyano cyclohexylidene acetic acid, acmc-20alci, cyclohexylidene cyanoacetic acid, cyclohexyliidenecyanoacetic acid, acetic acid, cyanocyclohexylidene, 2-cyano-2-cyclohexylideneaceticacid, cyano cyclohexylidene acetic acid #, acetic acid,2-cyano-2-cyclohexylidene
PubChem CID 97457
IUPAC Name 2-cyano-2-cyclohexylideneacetic acid
SMILES C1CCC(=C(C#N)C(=O)O)CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0167504 View Documents Alfa Aesar™
L0167504
2g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

37107-50-9
165.192
KMAMEGWTNNOKOE-UHFFFAOYSA-N
97457
C1CCC(=C(C#N)C(=O)O)CC1
C9H11NO2
MFCD00013799
cyclohexylidenecyanoacetic acid, cyanocyclohexylideneacetic acid, cyano cyclohexylidene acetic acid, acmc-20alci, cyclohexylidene cyanoacetic acid, cyclohexyliidenecyanoacetic acid, acetic acid, cyanocyclohexylidene, 2-cyano-2-cyclohexylideneaceticacid, cyano cyclohexylidene acetic acid #, acetic acid,2-cyano-2-cyclohexylidene
2-cyano-2-cyclohexylideneacetic acid

Specifications

107°C to 109°C
37107-50-9
MFCD00013799
1950877
KMAMEGWTNNOKOE-UHFFFAOYSA-N
2-cyano-2-cyclohexylideneacetic acid
97457
97%
2g
C9H11NO2
UN3439
cyclohexylidenecyanoacetic acid, cyanocyclohexylideneacetic acid, cyano cyclohexylidene acetic acid, acmc-20alci, cyclohexylidene cyanoacetic acid, cyclohexyliidenecyanoacetic acid, acetic acid, cyanocyclohexylidene, 2-cyano-2-cyclohexylideneaceticacid, cyano cyclohexylidene acetic acid #, acetic acid,2-cyano-2-cyclohexylidene
C1CCC(=C(C#N)C(=O)O)CC1
165.192
165.19
Cyclohexylidenecyanoacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.