missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexylhydrazine Hydrochloride, TCI America™

$177.22 - $516.41

Chemical Identifiers

CAS 24214-73-1
Molecular Formula C6H15ClN2
Molecular Weight (g/mol) 150.65
MDL Number MFCD00060160
InChI Key JZRHODNPRNTXKO-UHFFFAOYSA-N
Synonym cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine;
PubChem CID 207836
IUPAC Name cyclohexylhydrazine;hydrochloride
SMILES C1CCC(CC1)NN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C05005G View Documents TCI America
C05005G
5g Each for $177.22
 
C050025G View Documents TCI America
C050025G
25g Each for $516.41
 
Specifications

Chemical Identifiers

24214-73-1
150.65
JZRHODNPRNTXKO-UHFFFAOYSA-N
207836
C1CCC(CC1)NN.Cl
C6H15ClN2
MFCD00060160
cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine;
cyclohexylhydrazine;hydrochloride

Specifications

White-Yellow
5g
C6H15ClN2
cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine;
C1CCC(CC1)NN.Cl
150.65
150.65
≥98.0% (T)
112°C
24214-73-1
MFCD00060160
JZRHODNPRNTXKO-UHFFFAOYSA-N
cyclohexylhydrazine;hydrochloride
207836
Crystal-Powder at 20°C
Cyclohexylhydrazine Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No