Please login to your online account to display your discounted pricing

Cyclohexylhydrazine hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyclohexylhydrazine hydrochloride
Assay Percent Range 98%
Molecular Formula C6H14N2·ClH
Formula Weight 150.65
MDL Number MFCD00060160
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC430020250

  SDS
thermo fisher scientific
AC430020250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC430020050

  SDS
thermo fisher scientific
AC430020050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.65
Physical Form Crystalline Powder
Quantity 25g
Color White
Melting Point 110° to 115°C
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 46.64 to 49.03% C, 18.13 to 19.07% N
Chemical Name or Material Cyclohexylhydrazine hydrochloride
Molecular Formula C6H14N2·ClH
Assay Percent Range 98%
MDL Number MFCD00060160
CAS 24214-73-1
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.