missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexylhydrazine hydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 430020050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclohexylhydrazine hydrochloride | |
46.64 to 49.03% C, 18.13 to 19.07% N | |
98% | |
C6H14N2·ClH | |
5g | |
JZRHODNPRNTXKO-UHFFFAOYSA-N | |
cyclohexylhydrazine;hydrochloride | |
207836 | |
98% |
24214-73-1 | |
Authentic | |
Glass bottle | |
MFCD00060160 | |
cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine; | |
C1CCC(CC1)NN.Cl | |
150.65 | |
150.65 |
Chemical Identifiers
24214-73-1 | |
150.65 | |
JZRHODNPRNTXKO-UHFFFAOYSA-N | |
207836 | |
C1CCC(CC1)NN.Cl |
C6H14N2·ClH | |
MFCD00060160 | |
cyclohexylhydrazine hydrochloride, 1-cyclohexylhydrazine hydrochloride, cyclohexylhydrazinehydrochloride, hydrazine, cyclohexyl-, hydrochloride, 1-cyclohexylhydrazinehydrochloride, cyclohexyl hydrazine hydrochloride, hydrazine, cyclohexyl-, monohydrochloride, cyclohexyl-hydrazine hydrochloride, cyclohexyl-hydrazin;, cyclohexyl-hydrazine; | |
cyclohexylhydrazine;hydrochloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
RUO â Research Use Only