Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyldiphenylphosphine, 98%

$1,392.24 - $1,392.24

Chemical Identifiers

Chemical Name or Material Cyclohexyldiphenylphosphine
CAS 6372-42-5
Formula Weight 268.34
MDL Number MFCD00046360
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3670803

  SDS
alfa aesar™
3670803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3670806

  SDS
alfa aesar™
3670806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3670814

  SDS
alfa aesar™
3670814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3670822

  SDS
alfa aesar™
3670822
100g Each for $1,392.24
Description & Specifications

Specifications

Formula Weight 268.34
MDL Number MFCD00046360
Melting Point 58° to 62°C
Quantity 1g
CAS 6372-42-5
Sensitivity Air Sensitive
Chemical Name or Material Cyclohexyldiphenylphosphine
Assay 98%
Physical Form Solid
Flash Point >110° (230°F)