Please login to your online account to display your discounted pricing

Cyclohexylboronic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC359050010

 View more versions of this product

Catalog No. AC359050010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 127.97
Quantity 1g
Color Off-white
Chemical Name or Material Cyclohexylboronic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H13BO2
Linear Formula C6H11B(OH)2
MDL Number MFCD01311824
CAS 4441-56-9
Health Hazard 1 Warning
Recommended Storage Refrigerator +4°C
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclohexylboronic acid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H13BO2
Linear Formula C6H11B(OH)2
Formula Weight 127.97
MDL Number MFCD01311824
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Refrigerator +4°C

SDS & Safety Documents