missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine, 99%

CAS: 108-91-8 | C6H13N | 99.18 g/mol

Supplier:  Thermo Scientific Chemicals 111280025

 View more versions of this product

Catalog No. AC111280025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cyclohexylamine
-17.0°C
133.0°C to 134.0°C
Authentic
Glass bottle
1.4580 to 1.4600
2.5 L
0.8665
cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
PAFZNILMFXTMIY-UHFFFAOYSA-N
cyclohexanamine
7965
CHEBI:15773
99%
108-91-8
0.8665g/mL
27°C
98.5% min. (GC)
C6H13N
C6H11NH2
12,5
15,2722
Solubility in water: solublee. Other solubilities: miscible with most common organic solvents
C1CCC(CC1)N
99.18
2.4 mPa.s (20°C)
99.18
Liquid

Chemical Identifiers

108-91-8
99.18
cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
CHEBI:15773
C1CCC(CC1)N
C6H13N
PAFZNILMFXTMIY-UHFFFAOYSA-N
7965
cyclohexanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Suspected of damaging fertility.
Toxic in contact with skin.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr

GHS Signal Word: Danger

EINECSNumber : 203-629-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only