Learn More
Cyclohexylamine, 99%
CAS: 108-91-8 | C6H13N | 99.18 g/mol
Supplier: Thermo Scientific Chemicals 111280010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cyclohexylamine | |
| -17.0°C | |
| 133.0°C to 134.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4580 to 1.4600 | |
| 1 L | |
| 0.8665 | |
| cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro | |
| PAFZNILMFXTMIY-UHFFFAOYSA-N | |
| cyclohexanamine | |
| 7965 | |
| CHEBI:15773 | |
| 99% |
| 108-91-8 | |
| 0.8665g/mL | |
| 27°C | |
| 98.5% min. (GC) | |
| C6H13N | |
| C6H11NH2 | |
| 12,5 | |
| 15,2722 | |
| Solubility in water: solublee. Other solubilities: miscible with most common organic solvents | |
| C1CCC(CC1)N | |
| 99.18 | |
| 2.4 mPa.s (20°C) | |
| 99.18 | |
| Liquid |
Chemical Identifiers
| 108-91-8 | |
| 99.18 | |
| cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro | |
| CHEBI:15773 | |
| C1CCC(CC1)N |
| C6H13N | |
| PAFZNILMFXTMIY-UHFFFAOYSA-N | |
| 7965 | |
| cyclohexanamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Suspected of damaging fertility.
Toxic in contact with skin.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr
GHS Signal Word: Danger
EINECSNumber : 203-629-
RUO – Research Use Only