Learn More
Cyclohexylacetylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 380790050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclohexylacetylene | |
Yellow to Brown | |
130°C to 132°C | |
Authentic | |
Glass bottle | |
1.4530 to 1.455 | |
MFCD00001513 | |
0.828 | |
SSDZYLQUYMOSAK-UHFFFAOYSA-N | |
ethynylcyclohexane | |
70263 | |
98% |
931-48-6 | |
0.8280g/mL | |
18°C | |
97.5% min. (GC) | |
C8H12 | |
C6H11C∽CH | |
5 g | |
cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci | |
C#CC1CCCCC1 | |
108.18 | |
108.18 | |
Solution |
Chemical Identifiers
931-48-6 | |
108.18 | |
SSDZYLQUYMOSAK-UHFFFAOYSA-N | |
70263 | |
C#CC1CCCCC1 |
C8H12 | |
MFCD00001513 | |
cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci | |
ethynylcyclohexane |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Danger
EINECSNumber : 213-236-6
RUO – Research Use Only