missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexylacetylene, 98%

Manufacturer:  Alfa Aesar™ H5341814

 View more versions of this product

Catalog No. AAH5341814



Description

Description

Applications
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Cyclohexylacetylene
130°C to 132°C
931-48-6
MFCD00001513
1815535
SSDZYLQUYMOSAK-UHFFFAOYSA-N
ethynylcyclohexane
70263
98%
18°C (64°F)
25g
C8H12
UN3295
cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexylacetylene, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
C#CC1CCCCC1
108.184
108.18

Chemical Identifiers

931-48-6
108.184
SSDZYLQUYMOSAK-UHFFFAOYSA-N
70263
C#CC1CCCCC1
C8H12
MFCD00001513
cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexylacetylene, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
ethynylcyclohexane
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

SDS Documents