Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexylacetone, 97%

$139.07 - $1,735.78

Chemical Identifiers

Chemical Name or Material Cyclohexylacetone
CAS 103-78-6
Molecular Formula C9H16O
Formula Weight 140.23
MDL Number MFCD00019396
Beilstein 1209723
Synonym Acetonylcyclohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1274806

  SDS
alfa aesar™
A1274806
5g Each for $139.07

AAA1274822

  SDS
alfa aesar™
A1274822
100g Each for $1,735.78
Description & Specifications

Specifications

Formula Weight 140.23
Molecular Formula C9H16O
Quantity 5g
Beilstein 1209723
Boiling Point 198° to 200°C
Assay 97%
MDL Number MFCD00019396
Synonym Acetonylcyclohexane
Chemical Name or Material Cyclohexylacetone
TSCA TSCA
CAS 103-78-6