Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyl valerate, 97%

$69.66 - $227.64

Chemical Identifiers

Chemical Name or Material Cyclohexyl valerate
CAS 1551-43-5
Molecular Formula C11H20O2
Formula Weight 184.28g/mol
MDL Number MFCD00046355
Synonym Cyclohexyl pentanoate; Valeric acid cyclohexyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2165214

 
alfa aesar™
B21652-14
25g Each for $69.66

AAB2165222

 
alfa aesar™
B21652-22
100g Each for $227.64
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Cyclohexyl valerate
CAS 1551-43-5
Assay 97%
Molecular Formula C11H20O2
Formula Weight 184.28g/mol
MDL Number MFCD00046355
Synonym Cyclohexyl pentanoate; Valeric acid cyclohexyl ester