Learn More
Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04442604
Description
Cyclohexyl methyl methylphosphonate or phosphonic acid is used as a micro-nano electronic materials have extended self-assembled monolayers (SAMs) beyond conventional gold/thiol systems. To expand the choice of the substrate used for the preparation of SAMs, the chemical functionalities in film-forming molecules can be altered by introducing phosphate or phosphonate groups. Such polar acidic molecules are capable of interacting with diverse metal-oxide surfaces (e.g., Al2O3, Ta2O5, NbO5, ZrO2 and TiO2) and form films with a similar degree of ordering as for alkyl thiol SAMs on gold.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclohexyl methyl methylphosphonate | |
C8H17O3P | |
2 g | |
It is sparingly soluble in both organic solvents and water,but become more soluble in water when neutralized to phosphonates at neutral to high pH. | |
COP(=O)(C)OC1CCCCC1 | |
192.195 | |
192.09 |
7040-52-0 | |
MFCD08064211 | |
cyclohexyl methyl methylphosphonate, phosphonic acid, methyl-, cyclohexyl methyl ester, cyclohexyl methyl methyl-phosphonate, methoxy methyl phosphoryl oxycyclohexane, methylphosphonic acid methylcyclohexyl ester | |
UXITUBFQUAHYSL-UHFFFAOYSA-N | |
[methoxy(methyl)phosphoryl]oxycyclohexane | |
574009 | |
99% |
Chemical Identifiers
7040-52-0 | |
192.195 | |
UXITUBFQUAHYSL-UHFFFAOYSA-N | |
574009 | |
COP(=O)(C)OC1CCCCC1 |
C8H17O3P | |
MFCD08064211 | |
cyclohexyl methyl methylphosphonate, phosphonic acid, methyl-, cyclohexyl methyl ester, cyclohexyl methyl methyl-phosphonate, methoxy methyl phosphoryl oxycyclohexane, methylphosphonic acid methylcyclohexyl ester | |
[methoxy(methyl)phosphoryl]oxycyclohexane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only