Please login to your online account to display your discounted pricing

Cyclohexyl methacrylate, 97%, stabilized, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyclohexyl methacrylate
Name Note 97%
CAS 101-43-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H16O2
Linear Formula H2C=C(CH3)CO2C6H11
Formula Weight 168.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC310780050

  SDS
thermo fisher scientific
AC310780050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Cyclohexyl methacrylate
Color Clear colorless to slightly yellow
Quantity 5mL
Name Note 97%
CAS 101-43-9
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 202-943-5
TSCA TSCA
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Stabilizer 60 - 100ppm MEHQ
Physical Form Liquid
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H16O2
Linear Formula H2C=C(CH3)CO2C6H11
Formula Weight 168.24
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.