Please login to your online account to display your discounted pricing

Cyclohexyl methacrylate, 97%, stabilized, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC310780050

 View more versions of this product

Catalog No. AC310780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 168.24
Quantity 5mL
Color Clear colorless to slightly yellow
EINECS Number 202-943-5
TSCA TSCA
Infrared Spectrum Authentic
Name Note 97%
Stabilizer 60 - 100ppm MEHQ
Assay Percent Range 97%
CAS 101-43-9
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material Cyclohexyl methacrylate
Packaging Glass bottle
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Molecular Formula C10H16O2
Linear Formula H2C=C(CH3)CO2C6H11
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclohexyl methacrylate
Name Note 97%
CAS 101-43-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H16O2
Linear Formula H2C=C(CH3)CO2C6H11
Formula Weight 168.24
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents