Please login to your online account to display your discounted pricing

Cyclohexyl Mercaptan, 99+%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  111335000

 View more versions of this product

Catalog No. AC111335000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Liquid
Formula Weight 116.22
Quantity 500g
Color Clear
Beilstein 06, 8
TSCA TSCA
EINECS Number 216-378-7
RTECS Number GV7525000
Refractive Index 1.4915 to 1.4935 (20°C, 589nm)
Name Note 99+%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1569-69-3
Molecular Formula C6H12S
MDL Number MFCD00001455
Assay Percent Range 99+%
Linear Formula C6H11SH
Chemical Name or Material Cyclohexyl mercaptan
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym Cyclohexanethiol
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Flammable liquid and vapor.

Specifications

Chemical Name or Material Cyclohexyl mercaptan
Name Note 99+%
CAS 1569-69-3
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H12S
Linear Formula C6H11SH
Formula Weight 116.22
MDL Number MFCD00001455
Synonym Cyclohexanethiol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Flammable liquid and vapor.

SDS & Safety Documents