Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material Cyclohexyl isothiocyanate
CAS 1122-82-3
Formula Weight 141.24
MDL Number MFCD00003841
UN Number UN2922
Beilstein 774525
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1558506

 
alfa aesar™
A1558506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1558514

 
alfa aesar™
A1558514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.24
Chemical Name or Material Cyclohexyl isothiocyanate
TSCA TSCA
Odor Pungent
Refractive Index 1.5380
Physical Form Liquid
Assay 98%
MDL Number MFCD00003841
CAS 1122-82-3
UN Number UN2922
Beilstein 774525
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number NX8480000
Density 1.033g/mL
Sensitivity Moisture Sensitive
Boiling Point 219°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P308+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.