missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexyl Isocyanate, 98+%, ACROS Organics™

Manufacturer:  Acros Organics AC111310025

 View more versions of this product

Catalog No. AC111310025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclohexyl isocyanate
Authentic
1.4540 to 1.4560 (20°C, 589nm)
2.5kg
3173-53-3,95-50-1
C7H11NO
MFCD00003840
cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
C1CCC(CC1)N=C=O
125.171
CHEBI:136604
Liquid
0.9800g/mL
Glass bottle
Amber to Colorless
98+%
98+%
C6H11NCO
12, II, 12
KQWGXHWJMSMDJJ-UHFFFAOYSA-N
isocyanatocyclohexane
18502
125.17
≥98.0%

Chemical Identifiers

3173-53-3
125.171
KQWGXHWJMSMDJJ-UHFFFAOYSA-N
18502
isocyanatocyclohexane
C7H11NO
MFCD00003840
cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
CHEBI:136604
C1CCC(CC1)N=C=O
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor. Harmful in contact with skin. Harmful if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Very toxic to aquatic life. Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF exposed: Call a POISON CENTRE or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 221-639-3

RTECSNumber : NQ8650000

TSCA : TSCA

Recommended Storage : Product may develop some turbidity or precipitate

SDS Documents