missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexyl Isocyanate, 98+%, ACROS Organics™

$27.68 - $136.88

Chemical Identifiers

CAS 3173-53-3
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00003840
InChI Key KQWGXHWJMSMDJJ-UHFFFAOYSA-N
Synonym cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
PubChem CID 18502
ChEBI CHEBI:136604
IUPAC Name isocyanatocyclohexane
SMILES C1CCC(CC1)N=C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111310025 SDS View Documents Acros Organics
AC111310025
2.5kg Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC111310050 SDS View Documents Acros Organics
111310050
5g Glass bottle Each for $27.68
AC111315000 SDS View Documents Acros Organics
111315000
500g Glass bottle Each for $136.88
Specifications

Chemical Identifiers

3173-53-3
125.171
KQWGXHWJMSMDJJ-UHFFFAOYSA-N
18502
isocyanatocyclohexane
C7H11NO
MFCD00003840
cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
CHEBI:136604
C1CCC(CC1)N=C=O

Specifications

0.9800g/mL
Glass bottle
Amber to Colorless
3173-53-3,95-50-1
C7H11NO
MFCD00003840
cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
C1CCC(CC1)N=C=O
125.171
CHEBI:136604
Liquid
Cyclohexyl isocyanate, 98+%
Authentic
1.4540 to 1.4560 (20°C, 589nm)
2.5kg
98+%
C6H11NCO
12, II, 12
KQWGXHWJMSMDJJ-UHFFFAOYSA-N
isocyanatocyclohexane
18502
125.17
≥98.0%
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor. Harmful in contact with skin. Harmful if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Very toxic to aquatic life. Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF exposed: Call a POISON CENTRE or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 221-639-3

RTECSNumber : NQ8650000

TSCA : TSCA

Recommended Storage : Product may develop some turbidity or precipitate