Please login to your online account to display your discounted pricing

Cyclohexyl Isocyanate, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  111310050

 View more versions of this product

Catalog No. AC111310050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.0%
Formula Weight 125.17
Quantity 5g
Color Clear colorless to light amber
Beilstein 12, II, 12
EINECS Number 221-639-3
TSCA TSCA
RTECS Number NQ8650000
Chemical Name or Material Cyclohexyl isocyanate
Hydrolyzable Substance hydrolyzable chloride (titr.): < 0.3%
Infrared Spectrum Authentic
Packaging Glass bottle
Additional Information total other isocyanates (GC): < 2.0%
Name Note 98+%
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Storage Information Product may develop some turbidity or precipitate
Assay Percent Range 98+%
Molecular Formula C7H11NO
Linear Formula C6H11NCO
MDL Number MFCD00003840
CAS 3173-53-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very
CAS 95-50-1
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclohexyl isocyanate
Name Note 98+%
CAS 3173-53-3
CAS 95-50-1
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H11NO
Linear Formula C6H11NCO
Formula Weight 125.17
MDL Number MFCD00003840
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Very

SDS & Safety Documents