Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyl isocyanate, 98%

$27.04 - $27.04

Chemical Identifiers

Chemical Name or Material Cyclohexyl isocyanate
CAS 3173-53-3
Molecular Formula C7H11NO
Formula Weight 125.17g/mol
MDL Number MFCD00003840
UN Number UN2488
Beilstein 507983
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2305914

 
alfa aesar™
B23059-14
25g Each for $27.04

AAB2305922

 
alfa aesar™
B23059-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Pungent
Melting Point ≈−80°C
Boiling Point 168° to 170°C
Quantity 25g
Flash Point 48°C (118°F)
Chemical Name or Material Cyclohexyl isocyanate
CAS 3173-53-3
Assay 98%
Molecular Formula C7H11NO
Formula Weight 125.17g/mol
MDL Number MFCD00003840
UN Number UN2488
Beilstein 507983
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H330-H334-H318-H226-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYCLOHEXYL ISOCYANATE
RTECS Number NQ8650000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H330-H334-H318-H226-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Causes skin irritation.
May cause respiratory irritation.