missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexyl Diphenyl Phosphine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 406100050

 View more versions of this product

Catalog No. AC406100050


 

Specifications Description & Specifications

Specifications

Cyclohexyldiphenylphosphine
5g
Authentic
Powder
98%
268.33
White
6372-42-5
Glass bottle
≥97.5%
C18H21P
MFCD00046360

Chemical Identifiers

6372-42-5
C18H21P
MFCD00046360
≥97.5%
98%
268.33
Cyclohexyldiphenylphosphine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 228-904-2

RTECSNumber : SY8387500