Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyl(dimethoxy)methylsilane, 99%

Chemical Identifiers

Chemical Name or Material Cyclohexyl(dimethoxy)methylsilane
CAS 17865-32-6
Molecular Formula C9H20O2Si
Formula Weight 188.34
MDL Number MFCD00071752
UN Number UN3082
Beilstein 2827648
Synonym Cyclohexylmethyldimethoxysilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL16440AC

  SDS
alfa aesar™
L16440AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL16440AE

  SDS
alfa aesar™
L16440AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL16440AP

  SDS
alfa aesar™
L16440AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.34
Chemical Name or Material Cyclohexyl(dimethoxy)methylsilane
Molecular Formula C9H20O2Si
Health Hazard 1 Warning
Synonym Cyclohexylmethyldimethoxysilane
CAS 17865-32-6
Health Hazard 2 GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.
Assay 99%
TSCA TSCA
Flash Point 66°C (150°F)
Health Hazard 3 GHS P Statement
P210-P280-P362-P321-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 196°C
Beilstein 2827648
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Quantity 25mL
Melting Point <−72°C
UN Number UN3082
MDL Number MFCD00071752
Safety and Handling

Warning

GHS P Statement
P210-P280-P362-P321-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.