Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyl butyrate, 99%

$167.26 - $167.26

Chemical Identifiers

Chemical Name or Material Cyclohexyl butyrate
CAS 1551-44-6
Molecular Formula C10H18O2
Formula Weight 170.25
MDL Number MFCD00046354
Beilstein 2083414
Synonym Butyric acid cyclohexyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2069530

  SDS
alfa aesar™
B2069530
250g Each for $167.26

AAB2069518

  SDS
alfa aesar™
B2069518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Cyclohexyl butyrate
Boiling Point 90°C (15mmHg)
Quantity 250g
CAS 1551-44-6
RTECS Number ES8915000
TSCA TSCA
Assay 99%
Molecular Formula C10H18O2
Formula Weight 170.25
MDL Number MFCD00046354
Beilstein 2083414
Synonym Butyric acid cyclohexyl ester