missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexyl Bromide, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 159082500

 View more versions of this product

Catalog No. AC159082500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclohexyl bromide
1.3300g/mL
62°C
99%
C6H11Br
C6H11Br
250mL
1.33
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
AQNQQHJNRPDOQV-UHFFFAOYSA-N
bromocyclohexane
7960
99%
108-85-0
166.0°C to 167.0°C
Authentic
Glass bottle
1.4940 to 1.4960
MFCD00003819
05, 24
15, 2723
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
BrC1CCCCC1
163.06
163.06

Chemical Identifiers

108-85-0
163.06
AQNQQHJNRPDOQV-UHFFFAOYSA-N
7960
BrC1CCCCC1
C6H11Br
MFCD00003819
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n
bromocyclohexane
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
In use, may form flammable/explosive vapor-air mixture.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-622-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only