missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexyl Bromide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 159080050

 View more versions of this product

Catalog No. AC159080050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cyclohexyl bromide
62°C
Glass bottle
1.33
166.0°C to 167.0°C
5mL
99%
C6H11Br
05, 24
1-bromocyclohexane
AQNQQHJNRPDOQV-UHFFFAOYSA-N
bromocyclohexane
7960
Liquid
1.3300g/mL
Authentic
1.4940 to 1.4960
0.1% max. (K.F.)
Colorless to Yellow
108-85-0
C6H11Br
MFCD00003819
15, 2723
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
C1CCC(CC1)Br
163.06
163.06
99%

Chemical Identifiers

108-85-0
163.06
AQNQQHJNRPDOQV-UHFFFAOYSA-N
7960
C1CCC(CC1)Br
C6H11Br
MFCD00003819
1-bromocyclohexane
bromocyclohexane
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
In use, may form flammable/explosive vapor-air mixture.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-622-2

SDS Documents