missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol

Manufacturer:  Alfa Aesar™ L1292506*

 View more versions of this product

Catalog No. AAL1292506



Specifications

Specifications

Cyclohexyl acrylate
182°C to 184°C
stabilized with 100ppm 4-methoxyphenol
C9H14O2
UN3082
cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference
C=CC(=O)OC1CCCCC1
154.209
154.21
68°C (154°F)
5g
3066-71-5
MFCD00046349
1937528
KBLWLMPSVYBVDK-UHFFFAOYSA-N
cyclohexyl prop-2-enoate
18298
≥98%

Chemical Identifiers

3066-71-5
154.209
KBLWLMPSVYBVDK-UHFFFAOYSA-N
18298
C=CC(=O)OC1CCCCC1
C9H14O2
MFCD00046349
cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference
cyclohexyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H335-H227
Causes skin irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

RTECSNumber : AS7350000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents