Please login to your online account to display your discounted pricing

Alfa Aesar™ Cycloheximide, ultrapure, 98%

A protein synthesis inhibitor in eukaryotic cells

$67.19 - $67.19

Chemical Identifiers

Chemical Name or Material Cycloheximide
CAS 66-81-9
Name Note Ultrapure
Formula Weight 281.35
MDL Number MFCD00082346
UN Number UN2811
Beilstein 88868
Merck Index 14,2728
Synonym 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6690103

  SDS
alfa aesar™
J6690103
1g Each for $67.19
Description & Specifications

Specifications

Formula Weight 281.35
Melting Point 111° to 116°C
Assay 95%
Chemical Name or Material Cycloheximide
MDL Number MFCD00082346
CAS 66-81-9
Health Hazard 3 GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
UN Number UN2811
Merck Index 14,2728
Health Hazard 1 Danger
Boiling Point 492°C
Beilstein 88868
Synonym 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
Solubility Information Soluble in DMSO, acetone, ethanol and methanol; Slightly soluble in water with sonication
Health Hazard 2 GHS H Statement
H300-H360-H341
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Color White to cream
DOT Information Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number MA4375000
Name Note Ultrapure
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H360-H341
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.