Please login to your online account to display your discounted pricing

Alfa Aesar™ Cycloheximide solution,/mL in DMSO, 0.2 micron filtered

$124.36 - $124.36

Chemical Identifiers

Chemical Name or Material Cycloheximide solution
CAS 66-81-9
Name Note 100mg/mL in DMSO, 0.2 micron filtered
Formula Weight 281.35
MDL Number MFCD00082346
UN Number UN2810
Synonym CHX
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66004XF

  SDS
alfa aesar™
J66004XF
1mL Each for $124.36
Description & Specifications

Specifications

Formula Weight 281.35
Chemical Name or Material Cycloheximide solution
CAS 66-81-9
Health Hazard 2 GHS H Statement
H300-H360-H341-H227
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Combustible liquid.
Physical Form Liquid
UN Number UN2810
Synonym CHX
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P280-P281-P301+P310-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number MA4375000
Name Note 100mg/mL in DMSO, 0.2 micron filtered
MDL Number MFCD00082346
Color Colorless to pale yellow
Quantity 1mL
Safety and Handling

Danger

GHS P Statement
P210-P280-P281-P301+P310-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H360-H341-H227
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Combustible liquid.

2° to 8°C